Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
9003-01-4
C3H3NaO2
94.05
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
acrylic acid,2-propenoic acid,propenoic acid,vinylformic acid,acroleic acid,propene acid,ethylenecarboxylic acid,polyacrylate,propenoate,carbomer
6581
CHEBI:18308
prop-2-enoic acid
C=CC(=O)O
9003-01-4
C3H3NaO2
94.05
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
acrylic acid,2-propenoic acid,propenoic acid,vinylformic acid,acroleic acid,propene acid,ethylenecarboxylic acid,polyacrylate,propenoate,carbomer
6581
CHEBI:18308
prop-2-enoic acid
C=CC(=O)O
Linear Formula [-CH2CH(CO2H)-]n
Formula Weight 1.080.000 (Average )
Physical Form Powder
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Solubility Solubility in water: soluble.
Color White
Quantity 50g
Chemical Name or Material Poly(acrylic acid)

RTECSNumber : AT4680000

TSCA : TSCA