Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(caprolactone) Diol, Average M.W. 2000, ACROS Organics™

Click to view available options
Quantity:
250g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
36890-68-3
36890-68-3
Density 1.0700g/mL
Physical Form Waxy Solid
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in thf
Specific Gravity 1.07
Color White
Quantity 250g
Chemical Name or Material Poly(caprolactone) diol