Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(caprolactone) triol, average M.W. 300, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
39280-09-6
MFCD00084405
39280-09-6
MFCD00084405
Density 1.0700g/mL
Physical Form Viscous Liquid
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass Bottle
Refractive Index 1.4750 to 1.4780 (20°C, 589nm)
Color Colorless
Quantity 5g
Chemical Name or Material Poly(caprolactone) triol