Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(chloroprene), 10%, Acros Organics cis-85%, Acros Organics trans, Acros Organics

Click to view available options
Quantity:
250g
9010-98-4
C4H5Cl
88.534
MFCD00084406
YACLQRRMGMJLJV-UHFFFAOYSA-N
chloroprene,2-chloro-1,3-butadiene,chlorobutadiene,1,3-butadiene, 2-chloro,2-chlorobutadiene,neoprene,chloropreen,cloroprene,2-chlor-1,3-butadien,beta-chloroprene
31369
CHEBI:39481
2-chlorobuta-1,3-diene
C=CC(=C)Cl
9010-98-4
C4H5Cl
88.534
MFCD00084406
YACLQRRMGMJLJV-UHFFFAOYSA-N
chloroprene,2-chloro-1,3-butadiene,chlorobutadiene,1,3-butadiene, 2-chloro,2-chlorobutadiene,neoprene,chloropreen,cloroprene,2-chlor-1,3-butadien,beta-chloroprene
31369
CHEBI:39481
2-chlorobuta-1,3-diene
C=CC(=C)Cl
Chlorine (Cl) chlorine content: 40 wt%
Composition contains 10% cis and 85% trans
Linear Formula {-CH2CH=C(Cl)CH2-}n
Merck Index 10, 6308
Physical Form Chips or Chunks
Infrared Spectrum Authentic
Viscosity Mooney viscosity: 50
Color White to Beige
Quantity 250g
Chemical Name or Material Poly(chloroprene), 10% cis-85% trans