Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.W. 19,000, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
26780-50-7
C10H12O8
260.198
LCSKNASZPVZHEG-UHFFFAOYSA-N
vicryl,poly d,l-lactide-co-glycolide,polyglactin 370 usan,polyglactin,1,4-dioxane-2,5-dione-3,6-dimethyl-1,4-dioxane-2,5-dione 1:1,polyglactin 910 usan,vicryl rapide,vicryl plus
71391
3,6-dimethyl-1,4-dioxane-2,5-dione;1,4-dioxane-2,5-dione
CC1C(=O)OC(C(=O)O1)C.C1C(=O)OCC(=O)O1
26780-50-7
C10H12O8
260.198
LCSKNASZPVZHEG-UHFFFAOYSA-N
vicryl,poly d,l-lactide-co-glycolide,polyglactin 370 usan,polyglactin,1,4-dioxane-2,5-dione-3,6-dimethyl-1,4-dioxane-2,5-dione 1:1,polyglactin 910 usan,vicryl rapide,vicryl plus
71391
3,6-dimethyl-1,4-dioxane-2,5-dione;1,4-dioxane-2,5-dione
CC1C(=O)OC(C(=O)O1)C.C1C(=O)OCC(=O)O1
Physical Form Crystalline Powder or Granules
Packaging Glass bottle
Polymer 45 to 55% (Lactide), 45 to 55% (Glycolide)
Solubility Solubility in water: soluble.
Viscosity 0.15 to 0.25 dL/g
Color Tan to White
Quantity 5g
Chemical Name or Material Poly(DL-lactide-co-glycolide)