Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol), average M.W. 300, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
25322-68-3
MFCD01779599
PEG
25322-68-3
MFCD01779599
PEG
Appearance Clear liquid
Density 1.1200g/mL
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Hydroxyl Value 356 to 394mg KOH/g
Packaging Glass bottle
Refractive Index 1.4620 to 1.4650
Solubility Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Specific Gravity 1.12
Viscosity 85-100 mPa.s (20°C)
Color Index No. ≤50 APHA
Melting Point -15.0°C to +10.0°C
Quantity 1L
Flash Point >180°C
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

EINECSNumber : 500-038-2