Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol), average M.W. 400, ACROS Organics™

Click to view available options
Quantity:
1L
5L
Packaging:
Glass Bottle
Plastic drum
25322-68-3
MFCD01779601
PEG
25322-68-3
MFCD01779601
PEG
Appearance Clear liquid
Density 1.1275g/mL
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Hydroxyl Value 267 to 295mg KOH/g
Formula Weight 400
Physical Form Liquid
Packaging Glass Bottle
Refractive Index 1.4650 to 1.4680
Solubility Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Specific Gravity 1.1275
Viscosity 110-125 mPa.s (20°C)
Color Index No. ≤25 APHA
Boiling Point >200.0°C
Color Colorless
Melting Point 4.0°C to 8.0°C
Quantity 1L
Flash Point 235°C
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

EINECSNumber : 500-038-2