Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol) bis(3-aminopropyl) terminated, Thermo Scientific™

Click to view available options
Quantity:
1 g
5 g
34901-14-9
(C2H4O)nC6H16N2O
34901-14-9
(C2H4O)nC6H16N2O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 51.5°C to 54.0°C
Quantity 5 g
Chemical Name or Material Poly(ethylene glycol) bis(3-aminopropyl) terminated

GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

RUO – Research Use Only