Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly(ethylene glycol methyl ether), average M.W. 500, Acros Organics
Click to view available options
Quantity:
5g
500g
1kg
Packaging:
Glass bottle
Specifications
Melting Point | 12°C |
Viscosity | 20 mm2/s (50°C) |
Specific Gravity | 1.08 |
Flash Point | 268°C |
Density | 1.0800g/mL |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Sulfated Ash | 0.05% max. |
pH | 4.5 to 7.5 (5% aq. soln.) |
Water | 0.5% Max. |
Show More |