Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene glycol methyleether), average M.W. 750, ACROS Organics™

Click to view available options
Quantity:
5g
500g
1kg
Packaging:
Glass bottle
9004-74-4
C3H8O2
76.095
MFCD00084416
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve,ethanol, 2-methoxy,ethylene glycol monomethyl ether,methyl oxitol,2-methoxy-1-ethanol,methoxyethanol,3-oxa-1-butanol,egme,monomethyl glycol,dowanol em
8019
CHEBI:46790
2-methoxyethanol
COCCO
9004-74-4
C3H8O2
76.095
MFCD00084416
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve,ethanol, 2-methoxy,ethylene glycol monomethyl ether,methyl oxitol,2-methoxy-1-ethanol,methoxyethanol,3-oxa-1-butanol,egme,monomethyl glycol,dowanol em
8019
CHEBI:46790
2-methoxyethanol
COCCO
Additional Information diol-content: < 1 mole%
Appearance of Solution (25% aq. soln.) free from haze and turbidity
Avg. Mol. Wt. or Mol. Wt. Range 715 to 785
Density 1.09g/mL
Linear Formula CH3(OCH2CH)nOH
Ethylene Oxide < 20ppm
Hydroxyl Value 71.5 to 78.5mg KOH/g
Physical Form Fused Mass
Infrared Spectrum Authentic
Packaging Glass bottle
Sulfated Ash 0.05% max.
Water 0.1% max.
Color White
pH 4 to 7 (5% aq. soln.)
Quantity 5g
Chemical Name or Material Poly(ethylene glycol methyleether), Average M.W. 750
Show More Show Less

RTECSNumber : PE8263000

TSCA : TSCA