Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene oxide), 100%, ∽ M.W. 900,000, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
25322-68-3
MFCD00081839
PEO
25322-68-3
MFCD00081839
PEO
Linear Formula (-CH2CH2O-)n
Merck Index 15, 7688
Physical Form Powder
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Color White
Melting Point 65.0°C to 67.0°C
Quantity 5g
Chemical Name or Material Poly(ethylene oxide), Approximate M.W. 900, 000
Show More Show Less

RTECSNumber : TQ4100000

TSCA : TSCA