Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
poly-(LPAC) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$184.00 - $463.00
Specifications
Antigen | poly-(LPAC) |
---|---|
Applications | Immunohistochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Serum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN225825MI
|
MilliporeSigma
ABN225825UL |
25 μL |
Each for $184.00
|
|
|||||
ABN225810MI
|
MilliporeSigma
ABN2258100UL |
100 μL |
Each for $463.00
|
|
|||||
Description
poly-(LPAC) Polyclonal antibody specifically detects poly-(LPAC) in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Western Blotting.Specifications
poly-(LPAC) | |
Polyclonal | |
Serum | |
Neuroscience | |
(Leu-Pro-Ala-Cys)Exp | |
IgG | |
Unpurified | |
This rabbit polyclonal antibody specifically detects proteins with poly-(LPAC). |
Immunohistochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
A synthetic peptide corresponding to the (LPAC)EXP repeat motif. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼25 kDa observed. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title