Alfa Aesar™ Poly(methyl methacrylate)

Manufacturer: Alfa Aesar™ 43982A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43982A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9011-14-7
    Molecular Formula [-CH2C(CH3)(CO2CH3)-]n
    MDL Number MFCD00134349
    Chemical Name or Material Poly (methyl methacrylate)

    Specifications

    TSCA TSCA
    CAS 9011-14-7
    Color White
    Quantity 1kg
    Physical Form Powder
    Molecular Formula [-CH2C(CH3)(CO2CH3)-]n
    MDL Number MFCD00134349
    Melting Point >150°C
    RTECS Number TR0400000
    Chemical Name or Material Poly (methyl methacrylate)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit