Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(methyl methacrylate), ∽ M.W. 15000 (GPC), ACROS Organics™

Click to view available options
Quantity:
50g
100g
Packaging:
Plastic bottle
9011-14-7
MFCD00134349
Methyl methacrylate, polymerized,PMMA
9011-14-7
MFCD00134349
Methyl methacrylate, polymerized,PMMA
Density 1.18
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
Physical Form Beads
Infrared Spectrum Authentic
Loss on Drying 3% max.
Particle Size Bead size (typically): 10 to 100 micron
Packaging Plastic bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Color White
Quantity 50g
Flash Point >250°C
Chemical Name or Material Poly(methyl methacrylate), Average M.W. 15000 (GPC)
Show More Show Less

RTECSNumber : TR0400000

TSCA : TSCA