Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(methylhydrosiloxane), trimethylsilyl terminated, Acros Organics

Click to view available options
Quantity:
25mL
100mL
63148-57-2
63148-57-2
Infrared Spectrum Authentic
Density 1.0060g/mL
Flash Point 204°C
Packaging Glass bottle
Quantity 25mL
Specific Gravity 1.006
Linear Formula CH3H9OSi-(CH4OSi)n-CH3H9Si
Chemical Name or Material Poly(methylhydrosiloxane)