Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(N-vinylcarbazole), approx. M.W. 90,000, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
CAS 25067-59-8
Density 1.2000g/mL
Molecular Formula C14H11N
MDL Number MFCD00134336
Synonym 9-vinylcarbazole,n-vinylcarbazole,9-vinyl-9h-carbazole,9h-carbazole, 9-ethenyl,n-vinyl carbazole,vinylcarbazole,1-vinylcarbazole,poly 9-vinylcarbazole,carbazole, 9-vinyl,poly-n-vinylcarbazole
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
SMILES C=CN1C2=CC=CC=C2C3=CC=CC=C31
IUPAC Name 9-ethenylcarbazole
Molecular Weight (g/mol) 193.249
PubChem CID 15143
Physical Form Crystalline Powder
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Glass bottle
Color White
Quantity 25g
Chemical Name or Material Poly(N-vinylcarbazole), Secondary standard
Show More Show Less

RTECSNumber : FE6225480

TSCA : TSCA