Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly(phenylene sulfide), powder, nominal M.W. 10000, Thermo Scientific™
Click to view available options
5g; Glass bottle
Specifications
CAS | 9016-75-5 |
Linear Formula | (-C6H4S-)n |
MDL Number | MFCD00084445 |
Density | 1.3600g/mL |
Quantity | 5g |
Infrared Spectrum | Authentic |
Loss on Drying | 1% max. |
Packaging | Glass bottle |
Specific Gravity | 1.36 |
Chemical Name or Material | Poly(phenylene sulfide), nominal M.W. 10000 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction