Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(phenylene sulfide), powder, nominal M.W. 10000, ACROS Organics™

Click to view available options
Quantity:
5g
100g
250g
Packaging:
Glass bottle
Plastic bottle
9016-75-5
MFCD00084445
9016-75-5
MFCD00084445
Density 1.3600g/mL
Linear Formula (-C6H4S-)n
Physical Form Powder
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Plastic bottle
Specific Gravity 1.36
Color Tan to White
Quantity 100g
Chemical Name or Material Poly(phenylene sulfide), nominal M.W. 10000