Poly-Pro Inc Poly Sheeting

Full weight construction film

Manufacturer: Poly Pro Inc ASBPSBL0620

 View more versions of this product

Catalog No. 19-036-38

  Check Availability
Add to Cart

    Specifications

    Applications Construction, masonry and landscaping
    Color Black
    Certifications/Compliance Product Standard 17-69
    Thickness 6mil
    Product Type Poly Sheeting
    Material LLDPE/LDPE

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit