Poly-Pro Inc Poly Sheeting
Full weight construction film
Manufacturer: Poly Pro Inc PSCL0620
 View more versions of this product
Catalog No. 19-822-675
  Check Availability
Add to Cart
    Specifications
    Applications Jobsites, boats, insulation, vapor barrier, equipment cover, vapor barrier, insulation, paint protector or moisture barrier
    Color Clear
    Material LLDPE/LDPE
    Certifications/Compliance Product Standard 17-69
    Thickness 6mil
    Product Type Poly Sheeting
  • Description & Specifications
Related Products
Product Content Feedback