Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(propylene glycol), average M.W. 4.000, ACROS Organics™

Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
25322-69-4
C6H14O3
134.175
MFCD01779690
DUFKCOQISQKSAV-UHFFFAOYSA-N
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
32881
2-(2-hydroxypropoxy)propan-1-ol
CC(CO)OCC(C)O
25322-69-4
C6H14O3
134.175
MFCD01779690
DUFKCOQISQKSAV-UHFFFAOYSA-N
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
32881
2-(2-hydroxypropoxy)propan-1-ol
CC(CO)OCC(C)O
Density 1.0000g/mL
Linear Formula H[OCH(CH3)CH2]nOH
Physical Form Viscous Liquid
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4500 to 1.4520
Specific Gravity 1
Viscosity 930 mm2/s (25°C)
Color Green to Yellow
Quantity 5 g
Chemical Name or Material Poly(propylene glycol), Average M.W. 4.000
Show More Show Less

RTECSNumber : OE5784360

TSCA : TSCA

RMF - Research Use or Further Mfg