Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(propylene glycol), average M.W. 425, ACROS Organics™

Click to view available options
Quantity:
5g
250g
500g
2.5kg
Packaging:
Glass bottle
25322-69-4
C6H14O3
134.175
MFCD00084446
DUFKCOQISQKSAV-UHFFFAOYSA-N
polypropylene glycol,polyoxypropylene,2-2-hydroxypropoxy propan-1-ol,emkapyl,lineartop e,niax ppg,polyoxypropylene glycol,methyloxirane homopolymer,desmophen 360c,2-2-hydroxypropoxy-1-propanol
32881
2-(2-hydroxypropoxy)propan-1-ol
CC(CO)OCC(C)O
25322-69-4
C6H14O3
134.175
MFCD00084446
DUFKCOQISQKSAV-UHFFFAOYSA-N
polypropylene glycol,polyoxypropylene,2-2-hydroxypropoxy propan-1-ol,emkapyl,lineartop e,niax ppg,polyoxypropylene glycol,methyloxirane homopolymer,desmophen 360c,2-2-hydroxypropoxy-1-propanol
32881
2-(2-hydroxypropoxy)propan-1-ol
CC(CO)OCC(C)O
Density 1.0000g/mL
Linear Formula H[OCH(CH3)CH2]nOH
Packaging Glass bottle
Refractive Index 1.4465 to 1.4505
Solubility Solubility in water: completely soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, chloroform, dioxane, ether, thf
Specific Gravity 1
Viscosity 80 mm2/s (25°C)
Boiling Point 200.0°C
Melting Point -25.0°C
Quantity 250g
Flash Point 185°C
Chemical Name or Material Poly(propylene glycol), Average M.W. 425
Show More Show Less

RTECSNumber : OE5784360

TSCA : TSCA