Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Poly(propylene glycol) bis(2-aminopropyl ether), Mn ca 230

Manufacturer:  Alfa Aesar™ H37543AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH37543AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

9046-10-0
UN2922
JEFFAMINE|r D-230
Poly(propylene glycol) bis(2-aminopropyl ether), Mn ≈230

Specifications

9046-10-0
UN2922
0.948g/mL
500mL
Liquid
JEFFAMINE|r D-230
colorless to pale yellow
Poly(propylene glycol) bis(2-aminopropyl ether), Mn ≈230

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : TR3702500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit