Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(styrene), average M.W. 250,000, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
1kg
Packaging:
Glass bottle
9003-53-6
C8H8
104.152
MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
styrene
C=CC1=CC=CC=C1
9003-53-6
C8H8
104.152
MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
styrene
C=CC1=CC=CC=C1
Density 1.0500g/mL
Linear Formula [-CH2CH(C6H5)-]n
Merck Index 14, 8860
Physical Form Pellets
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Refractive Index 1.5916
Solubility Solubility in water: insoluble. Other solubilities: soluble in ethylbenzene, methyl isobutyl ketone,, thf, benzene, toluene, pyridine, methylenechloride
Specific Gravity 1.05
Color Undesignated
Melting Point 93.0°C
Quantity 25g
Chemical Name or Material Poly(styrene)
Show More Show Less