Alfa Aesar™ Poly(styrene-divinylbenzene), 2% cross-linked, 200-400 mesh

Preparation of Merrifield type resins

Manufacturer: Alfa Aesar™ 4470822

Quantity 100g

 View more versions of this product

Catalog No. AA4470822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9003-70-7
    MDL Number MFCD00146442
    Chemical Name or Material Poly (styrene-divinylbenzene), 2% cross-linked

    Specifications

    MDL Number MFCD00146442
    CAS 9003-70-7
    Mesh Size 200 to 400 Mesh
    Odor Odorless
    Quantity 100g
    TSCA TSCA
    Physical Form Powder
    Chemical Name or Material Poly (styrene-divinylbenzene), 2% cross-linked

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit