Alfa Aesar™ Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 200-400 mesh

Manufacturer: Alfa Aesar™ 4439422

Quantity 100g

 View more versions of this product

Catalog No. AA4439422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89551-24-6
    MDL Number MFCD00146442
    Synonym Aminomethyl polystyrene
    Chemical Name or Material Poly (styrene-divinylbenzene), Aminomethylated, 1% cross-linked

    Specifications

    CAS 89551-24-6
    MDL Number MFCD00146442
    Synonym Aminomethyl polystyrene
    Quantity 100g
    Physical Form Powder
    Mesh Size 200 to 400 Mesh
    Chemical Name or Material Poly (styrene-divinylbenzene), Aminomethylated, 1% cross-linked

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit