Alfa Aesar™ Poly(styrene sulfonic acid) sodium salt, M.W. 300,000

Manufacturer: Alfa Aesar™ 4504914

Quantity 25g

 View more versions of this product

Catalog No. AA4504914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25704-18-1
    Molecular Formula [-CH2CH(C6H4)SO3Na)-]n
    MDL Number MFCD00084449
    Chemical Name or Material Poly (styrene sulfonic acid) sodium salt, M.W. 300,000

    Specifications

    Physical Form Powder
    Molecular Formula [-CH2CH(C6H4)SO3Na)-]n
    CAS 25704-18-1
    Quantity 25g
    MDL Number MFCD00084449
    Color Pale brown
    Chemical Name or Material Poly (styrene sulfonic acid) sodium salt, M.W. 300,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit