Alfa Aesar™ Poly(styrene sulfonic acid) sodium salt, M.W. 75,000

Manufacturer: Alfa Aesar™ 4585118

Quantity 50g

 View more versions of this product

Catalog No. AA4585118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25704-18-1
    Molecular Formula [-CH2CH(C6H4)SO3Na)-]n
    MDL Number MFCD00084449
    Chemical Name or Material Poly (styrene sulfonic acid) sodium salt, M.W. 75,000

    Specifications

    CAS 25704-18-1
    TSCA TSCA
    Physical Form Powder
    Color Pale brown
    Molecular Formula [-CH2CH(C6H4)SO3Na)-]n
    MDL Number MFCD00084449
    Quantity 50g
    RTECS Number DB8550000
    Chemical Name or Material Poly (styrene sulfonic acid) sodium salt, M.W. 75,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit