Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl acetate), ∽ M.W. 170,000, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
Packaging:
Glass bottle
9003-20-7
C4H6O2
86.09
MFCD00084457
XTXRWKRVRITETP-UHFFFAOYSA-N
vinyl acetate,acetic acid ethenyl ester,acetic acid vinyl ester,ethenyl ethanoate,1-acetoxyethylene,vinyl ethanoate,acetoxyethene,vinylacetat,vinyl acetate monomer,vinyl a monomer
7904
CHEBI:46916
ethenyl acetate
CC(=O)OC=C
9003-20-7
C4H6O2
86.09
MFCD00084457
XTXRWKRVRITETP-UHFFFAOYSA-N
vinyl acetate,acetic acid ethenyl ester,acetic acid vinyl ester,ethenyl ethanoate,1-acetoxyethylene,vinyl ethanoate,acetoxyethene,vinylacetat,vinyl acetate monomer,vinyl a monomer
7904
CHEBI:46916
ethenyl acetate
CC(=O)OC=C
Density 1.1800g/mL
Linear Formula [-CH2CH(O2CCH3)-]n
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in low molecular weight alcohols, esters,, benzene and chorinated hydrocarbons
Physical Form Beads
Infrared Spectrum Authentic
Loss on Drying 4% max.
Packaging Glass bottle
Specific Gravity 1.18
Color Undesignated
Quantity 1kg
Chemical Name or Material Poly(vinyl acetate), Average M.W. 170.000
Show More Show Less

RTECSNumber : AK0920000

TSCA : TSCA