Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl alcohol), 95% hydrolyzed, average M.W. 95000, ACROS Organics™

Click to view available options
Quantity:
500g
Indicator
CAS 9002-89-5
Ash 0.9% max.
Molecular Formula C2H4O
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Merck Index 15, 7706
Synonym vinyl alcohol,polyvinyl alcohol,hydroxyethene,hydroxyethylene,ethenol, homopolymer,gohsenol,polyviol,elvanol,mowiol,poval
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES C=CO
IUPAC Name ethenol
Molecular Weight (g/mol) 44.053
PubChem CID 11199
Physical Form Granular Powder
Infrared Spectrum Authentic
Percent Purity 95.5 to 96.5 mole% (degree of hydrolysis)
Loss on Drying 5% max.
Packaging Glass bottle
Viscosity 28 to 31 mPa·s (20°C, 4% aq. sol.)
Color White to Cream
Quantity 500g
Chemical Name or Material Poly(vinyl alcohol), 95% Hydrolyzed, Average M.W. 95000
Show More Show Less

RTECSNumber : TR8100000

TSCA : TSCA