Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
9002-89-5
C2H4O
44.053
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol,polyvinyl alcohol,hydroxyethene,hydroxyethylene,ethenol, homopolymer,gohsenol,polyviol,elvanol,mowiol,poval
11199
ethenol
C=CO
9002-89-5
C2H4O
44.053
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol,polyvinyl alcohol,hydroxyethene,hydroxyethylene,ethenol, homopolymer,gohsenol,polyviol,elvanol,mowiol,poval
11199
ethenol
C=CO
Indicator
Ash 1% max.
Linear Formula [-CH2CH(OH)-]n
Merck Index 15, 7706
Physical Form Powder or Granules
Infrared Spectrum Authentic
Percent Purity 98 mole% min. (degree of hydrolysis)
Loss on Drying 5% max.
Packaging Glass bottle
Viscosity 6mPa·s (20°C 4% aq. solution)
Color White
Quantity 100g
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less