Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl alcohol), 98.0-98.8% hydrolyzed,M.W. approx. 146,000-186,000, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
9002-89-5
C2H4O
44.053
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol,polyvinyl alcohol,hydroxyethene,hydroxyethylene,ethenol, homopolymer,gohsenol,polyviol,elvanol,mowiol,poval
11199
ethenol
C=CO
9002-89-5
C2H4O
44.053
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol,polyvinyl alcohol,hydroxyethene,hydroxyethylene,ethenol, homopolymer,gohsenol,polyviol,elvanol,mowiol,poval
11199
ethenol
C=CO

Indicator

Quantity 100g
pH 5.0 to 7.0 (4% aq. soln.)
Melting Point 230.0°C to 240.0°C
Infrared Spectrum Authentic
Viscosity 62-72 mPa.s (20°C 4% aq. sol.)
Linear Formula [-CH2CH(OH)-]n
Merck Index 15,7706
Solubility Information Solubility in water: soluble in hot water.
Percent Purity 98.0-98.8%
Chemical Name or Material Poly(vinyl alcohol)