Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
Packaging:
Glass bottle
25702-80-1
C5H7ClO2
134.559
MFCD00084460
SQNNHEYXAJPPKH-UHFFFAOYSA-N
2-propenoic acid, polymer with chloroethene,polyvinyl chloride-acrylic copolymer,acrylic acid; chloroethylene,chloroethene; prop-2-enoic acid,acrylic acid; vinyl chloride
3080734
chloroethene;prop-2-enoic acid
C=CC(=O)O.C=CCl
25702-80-1
C5H7ClO2
134.559
MFCD00084460
SQNNHEYXAJPPKH-UHFFFAOYSA-N
2-propenoic acid, polymer with chloroethene,polyvinyl chloride-acrylic copolymer,acrylic acid; chloroethylene,chloroethene; prop-2-enoic acid,acrylic acid; vinyl chloride
3080734
chloroethene;prop-2-enoic acid
C=CC(=O)O.C=CCl
Density 1.3900g/mL
Physical Form Amorphous Powder
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in acetone, carbon disulfide, mek, thf
Specific Gravity 1.39
Color White
Quantity 100g
Chemical Name or Material Poly(vinyl chloride), carboxylated