Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl ethyleether), low molecular weight, ACROS Organics™

Click to view available options
Quantity:
100g
250g
25104-37-4
C4H8O
72.107
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethyl vinyl ether,ethoxyethene,ethoxyethylene,ethene, ethoxy,1-ethoxyethene,1-ethoxyethylene,vinyl ethyl ether,vinamar,ether, ethyl vinyl,ether, vinyl ethyl
8023
ethenoxyethane
CCOC=C
25104-37-4
C4H8O
72.107
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethyl vinyl ether,ethoxyethene,ethoxyethylene,ethene, ethoxy,1-ethoxyethene,1-ethoxyethylene,vinyl ethyl ether,vinamar,ether, ethyl vinyl,ether, vinyl ethyl
8023
ethenoxyethane
CCOC=C
Density 96%g/mL
Linear Formula {-CH2CH(OC2H5)-}n
Physical Form Viscous Liquid
Infrared Spectrum Authentic
Loss on Drying 10% max.
Color Amber
Quantity 100g
Chemical Name or Material Poly(vinyl ethyleether)

RTECSNumber : CQ6693000

TSCA : TSCA