Alfa Aesar™ Poly(vinyl formal)

Manufacturer: Alfa Aesar™ 4419014

Quantity 25g

 View more versions of this product

Catalog No. AA4419014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9003-33-2
    MDL Number MFCD00084463
    Chemical Name or Material Poly (vinyl formal)

    Specifications

    Physical Form Granular Powder
    Density 1.23g/mL
    MDL Number MFCD00084463
    CAS 9003-33-2
    TSCA TSCA
    Solubility Information Soluble in acetone, chloroform, dioxane, THF
    Quantity 25g
    Chemical Name or Material Poly (vinyl formal)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit