Alfa Aesar™ Poly(vinylidene chloride-co-acrylonitrile), 80/20

Manufacturer: Alfa Aesar™ 4375736

Quantity 500g

 View more versions of this product

Catalog No. AA4375736

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 9010-76-8
    Molecular Formula (-CH2CCl2-)x[-CH2CH(CN)-]y
    MDL Number MFCD00134022
    Chemical Name or Material Poly (vinylidene chloride-co-acrylonitrile), 80/20

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Physical Form Powder
    Molecular Formula (-CH2CCl2-)x[-CH2CH(CN)-]y
    MDL Number MFCD00134022
    CAS 9010-76-8
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Density 1.60g/mL
    Color Off-white
    Quantity 500g
    Chemical Name or Material Poly (vinylidene chloride-co-acrylonitrile), 80/20

    GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit