Alfa Aesar™ Poly(vinylidene chloride-co-acrylonitrile), 80/20

Manufacturer: Alfa Aesar™ 4375722

Quantity 100g

 View more versions of this product

Catalog No. AA4375722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9010-76-8
    Molecular Formula (-CH2CCl2-)x[-CH2CH(CN)-]y
    MDL Number MFCD00134022
    Chemical Name or Material Poly (vinylidene chloride-co-acrylonitrile), 80/20

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Off-white
    Density 1.60g/mL
    Molecular Formula (-CH2CCl2-)x[-CH2CH(CN)-]y
    MDL Number MFCD00134022
    Quantity 100g
    CAS 9010-76-8
    Chemical Name or Material Poly (vinylidene chloride-co-acrylonitrile), 80/20

    GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit