Alfa Aesar™ Polyaniline, emeraldine base

Conductive polymer

Manufacturer: Alfa Aesar™ 4258114

Quantity 25g

 View more versions of this product

Catalog No. AA4258114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25233-30-1
    MDL Number MFCD00241446
    Chemical Name or Material Polyaniline, Emeraldine base

    Specifications

    Physical Form Powder
    MDL Number MFCD00241446
    Color Dark brown
    Melting Point >200°C (dec.)
    Quantity 25g
    CAS 25233-30-1
    Chemical Name or Material Polyaniline, Emeraldine base

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit