Alfa Aesar™ Polyaniline, emeraldine salt from p-toluenesulfonic acid

Manufacturer: Alfa Aesar™ 4368330

Quantity 250g

 View more versions of this product

Catalog No. AA4368330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25233-30-1
    MDL Number MFCD00284320
    Chemical Name or Material Polyaniline, Emeraldine salt from p-toluenesulfonic acid

    Specifications

    Physical Form Powder
    CAS 25233-30-1
    Color Dark green
    MDL Number MFCD00284320
    Melting Point >300°C (dec.)
    Quantity 250g
    Chemical Name or Material Polyaniline, Emeraldine salt from p-toluenesulfonic acid

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit