Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega PolyATtract™ mRNA Isolation Systems
Click to view available options
Quantity:
15 Preps
Includes:
mRNA Isolation System only
mRNA Isolation System, stand
Description
- Isolates Highly Pure mRNA from Total RNA
- Protocol complete in approximately 45 minutes
- Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
- Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
- Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000μg total RNA
Specifications
Specifications
Content And Storage | 2°C to 10°C; Nuclease-free Water 2 x 25mL, SSC (20X) 2 x 1.4mL, Biotinylated Oligo(dT) probe 1 x 50μL, Streptavidin MagneSphere™ paramagnetic particles 15 x 0.6mL |
For Use With (Application) | In-vitro translation, RT-PCR and cDNA synthesis |
Includes | mRNA Isolation System only |
Quantity | 15 Preps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction