Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polycarbonate resin, ∽ M.W. 45.000, pellets, Thermo Scientific Chemicals

Catalog Number p-185828
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
24936-68-3
(C16H14O3)n
NaN
MFCD00084476
CC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1
24936-68-3
(C16H14O3)n
NaN
MFCD00084476
CC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Undesignated
Physical Form Pellets
Packaging Glass bottle
Density 1.2000g/mL
Quantity 5 g
Linear Formula [-C6H4C(CH3)2C6H4OCO2-]n
Chemical Name or Material Polycarbonate resin, M.W. 64.000
RTECSNumber TR1580150

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit