Learn More
Cell Signaling TechnologySupplier Diversity Partner Phospho-Elk-1 (Ser383) Antibody 100 ul DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 9181S
9181S , The transcription factor Elk-1 is a component of the ternary complex that binds the serum response element (SRE) and mediates gene activity in response to serum and growth factors (1-3). Elk-1 is phosphorylated by MAP kinase pathways at a cluster of S/T motifs at its carboxy terminus; phosphorylation at these sites, particularly Ser383, is critical for transcriptional activation by Elk-1. Elk-1 appears to be a direct target of activated MAP kinase: (a) biochemical studies indicate that Elk-1 is a good substrate for MAP kinase; (b) the kinetics of Elk-1 phosphorylation and activation correlate with MAP kinase activity; (c) interfering mutants of MAP kinase block Elk-1 activation in vivo. Other studies have shown that Elk-1 (Ser383) is also a target of the stress-activated kinase SAPK/JNK (4,5).
Catalog No. 50-194-767