Learn More
Cell Signaling TechnologySupplier Diversity Partner Phospho-MYPT1 (Thr696) Antibody 100 ul DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 5163S
5163S , Protein phosphatase 1 (PP1) is a ubiquitous eukaryotic protein serine/threonine phosphatase involved in the regulation of various cell functions. Substrate specificity is determined by the binding of a regulatory subunit to the PP1 catalytic subunit (PP1c). It is estimated that over fifty different regulatory subunits exist (1).
Catalog No. 50-194-694