Learn More
Cell Signaling TechnologySupplier Diversity Partner Ran Antibody 100 ul DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 4462S
4462S , Ran is a small GTPase of the Ras family that plays a central role in the spacial and temperal organization of eukaryotic cells. During interphase, Ran-GDP localizes to the cytoplasm and Ran-GTP to the nucleus. This polarized localization of Ran ensures its role in nuclear transport (1). During mitosis, Ran-GTP is chromatin associated, where it promotes spindle assembly and nuclear envolope formation (1,2). In S phase, Ran-GTP associates with and inhibits MCM helicase, ensuring precise chromosomal DNA duplication during the cell cycle (3).
Catalog No. 50-195-118