Learn More
Cell Signaling TechnologySupplier Diversity Partner Ras-GRF1 Antibody 100 ul DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 3322S
3322S , Ras activity is regulated by GAP (GTPase activating proteins) and GEFs (guanine nucleotide exchange factors). Ras-GRF1 (also known as CDC25Mm) is neuronal RasGEF and is regulated by heterotrimeric G proteins and calcium influx (1,2). Binding to calmodulin and phosphorylation stimulate Ras-GRF1 activity (1,2). Multiple PKA phosphorylation sites on Ras-GRF have been identified. Phosphorylation on the two major sites, Ser54 and Ser822, inhibits Ras-GRF activity (3). Carbachol (a muscarinic agonist)-induced phosphorylation on Ser916 is essential but not sufficient for maximal Ras-GRF activity (4). It has been reported that Ras-GRF1 also shows GEF activity toward Rac after phosphorylation by the tyrosine kinase Src (5).
Catalog No. 50-195-124