PolyCo™ VR™ Protective Boot Covers

Provide knee-high, foot and lower leg protection in situations where biological and chemical liquids are present

Manufacturer: PolyConversions, Inc. 49555

 View more versions of this product

Catalog No. 19-125-546

  Check Availability
Add to Cart

    Specifications

    Size Large
    Product Type Protective Boot Covers
    Material PolyCo VR™, Polyolefin
    For Use With (Application) Ideal for use where biological and chemical liquids are present
    Height (English) 14 in.
    Closure Type Elastic
    Height (Metric) 35.5cm
    Seam Type Heat sealed
    Thickness 6mil
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit