Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PolyCo™ VR™ Protective Gowns with Thumb Loops
Description
- Glove-to-sleeve protection
- Full-frontal coverage suitable for workplaces with biological or chemical hazards
- Slips on over workers' heads and ties securely in the back for a quick and easy fit
- Secures sleeves in place with thumb loop
- Durable 4 mil thickness
Specifications
Length (English) | 45 in. |
Length (Metric) | 114.3 cm |
Product Type | Controlled Environments Gown |
For Use With (Application) | Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing |
Closure Type | Ties |
Sterility | Non-sterile |
Seam Type | Welded |
Sleeve Style | Full Sleeve with Thumb Loop |
Certifications/Compliance | OSHA 29, CFR Part 1910.1030 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction