Learn More
Polycystin-2 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: YCE2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma MABS2145100UG
 View more versions of this product
Catalog No. MABS21451MI
Add to cart
Description
Description
Polycystin-2 Monoclonal antibody specifically detects Polycystin-2 in Human,Mouse,Rat samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications
Specifications
Polycystin-2
YCE2
NP_000288
PC2;Autosomal dominant polycystic kidney disease type II protein;Polycystic kidney disease 2 protein;Polycystwin;R48321;Transient receptor potential cation channel subfamily P member 2
Mouse
IgG2aκ
Protein G Purified
Signaling
Primary
Clone YCE2 is a mouse monoclonal antibody that specifically detects Polycystin-2. It targets an epitope within 68 amino acids from the cytoplasmic domain in the C-terminal region.
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Purified
PKD2, TRPP2
GST-tagged recombinant fragment corresponding to 68 amino acids from the C-terminal region, cytoplasmic domain.
109.69 kDa calculated.
100 μg
Stable for 1 year at 2 to 8°C from date of receipt.
Monoclonal
Human, Mouse, Rat