Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polycystin-2 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: YCE2, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $444.00
Specifications
Antigen | Polycystin-2 |
---|---|
Clone | YCE2 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS21452MI
|
MilliporeSigma
MABS214525UG |
25 μg |
Each for $177.00
|
|
|||||
MABS21451MI
|
MilliporeSigma
MABS2145100UG |
100 μg |
Each for $444.00
|
|
|||||
Description
Polycystin-2 Monoclonal antibody specifically detects Polycystin-2 in Human,Mouse,Rat samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.Specifications
Polycystin-2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
PC2;Autosomal dominant polycystic kidney disease type II protein;Polycystic kidney disease 2 protein;Polycystwin;R48321;Transient receptor potential cation channel subfamily P member 2 | |
GST-tagged recombinant fragment corresponding to 68 amino acids from the C-terminal region, cytoplasmic domain. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
109.69 kDa calculated. |
YCE2 | |
Monoclonal | |
Purified | |
Signaling | |
NP_000288 | |
PKD2, TRPP2 | |
IgG2a κ | |
Protein G Purified | |
Clone YCE2 is a mouse monoclonal antibody that specifically detects Polycystin-2. It targets an epitope within 68 amino acids from the cytoplasmic domain in the C-terminal region. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title