Learn More
Medchemexpress LLC Polydatin | 27208-80-6 | 99.50% | 390.38 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN0120A10MM/1ML
Encompass_Preferred
Polydatin (Piceid), extracted from the roots of Reynoutria japonica, a widely used traditional Chinese remedy, possesses anti-inflammatory activity in several experimental models. It inhibits G6PD and induces oxidative and ER stresses. It is for research use only.
  • Protects cerebral cells from ischemic damages via improvement of microcirculation and inhibition of platelet aggregation.
  • Inhibits ICAM-1 expression in endothelial cells stimulated by lipopolysaccharide, and attenuates adhesion between white blood cells and endothelial cells.
  • Inhibits cancer cell proliferation and cell cycle progression.
  • Induces ER stress-driven cell death.
  • Increases the activity of SOD and the heart rate, attenuates myocardial pathological damage, decreases MDA content, slightly increases arterial pressure and GSH-Px activity, reduces intervals of QRS, QT and ST, and lowers FFA content.
Catalog No. 50-004-30280